accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3

CAS RN 97-87-0

IUPAC Name: butyl 2-methylpropanoate
Synonyms: butyl isobutyrate isobutyric acid, butyl ester (6CI,7CI,8CI)
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: JSLCOZYBKYHZNL-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results