accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97-93-8

IUPAC Name: triethylalumane
Synonyms: triethylaluminium aluminum triethyl
Molecular Weight (g/mol): 114.17
Molecular Formula: C6H15Al
InChi Key: VOITXYVAKOUIBA-UHFFFAOYSA-N
SMILES: CC[Al](CC)CC
Molecular Weight (g/mol) 
  • (7)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (1)
  • (4)
  • (7)
  • (3)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)