Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylaluminum (15% in Toluene, ca. 1.1mol/L), TCI America™
SDP

Catalog No. T0925100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0925100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0925100ML Supplier TCI America Supplier No. T0925100ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97-93-8
Molecular Formula C6H15Al
Molecular Weight (g/mol) 114.168
MDL Number MFCD00009015
InChI Key VOITXYVAKOUIBA-UHFFFAOYSA-N
Synonym triethylaluminum, triethylaluminium, aluminum, triethyl, triethylalane, unii-h426e9h3tt, triethylaluminum solution, 1.0 m in hexanes, triethylaluminum solution, 25 wt. % in toluene, triethyl-aluminum, triethyl aluminum, triethyl-aluminum
PubChem CID 16682930
IUPAC Name triethylalumane
SMILES CC[Al](CC)CC

Specifications

Color Colorless
UN Number 3394
Quantity 100 mL
Formula Weight 114.17
Physical Form Liquid
Chemical Name or Material Triethylaluminum (15% in Toluene, ca. 1.1mol/L)
EINECSNumber (2)-2227
RTECSNumber BD2050000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.