CAS RN 97075-46-2
IUPAC Name:
(diphenylmethyl)({2-[(diphenylmethyl)amino]ethyl})amine dihydrochloride
Synonyms:
(diphenylmethyl)({2-[(diphenylmethyl)amino]ethyl})amine dihydrochloride
Molecular Weight (g/mol):
465.46
Molecular Formula:
C28H30Cl2N2
InChi Key:
YRQCDCNQANSUPB-UHFFFAOYSA-N
SMILES:
Cl.Cl.C(CNC(C1=CC=CC=C1)C1=CC=CC=C1)NC(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ AMN 082 dihydrochloride
CAS: 97075-46-2 Molecular Formula: C28H30Cl2N2