accessibility menu, dialog, popup

UserName

Viewing profile as user:
FePPCH3H3CCH3CH3H3CCH3H3CCH3

CAS RN 97239-80-0

IUPAC Name: 1,1'-Bis(diisopropylphosphino)ferrocene
Synonyms:
Molecular Weight (g/mol): 418.32
Molecular Formula: C22H36FeP2
InChi Key:
SMILES: [Fe].CC(C)P(C(C)C)c1cccc1.CC(C)P(C(C)C)c1cccc1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)