Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Bis(diisopropylphosphino)ferrocene, 98%, Thermo Scientific Chemicals

Catalog No. AAH2705706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2705706 5 g
AAH2705703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2705706 Supplier Thermo Scientific Chemicals Supplier No. H2705706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

The general and efficient cross-coupling of thiols with aryl halides was developed utilizing Pd(OAc) 2 /1,1-bis(diisopropylphosphino)ferrocene as the catalyst. The oxidative electrochemistry of dippf was examined, and it was found to follow an ECDim mechanism. Seven new compounds containing dippf were prepared and characterized spectroscopically. The crystal structures of [MCl2(dippf)] (M = Pt and Zn) were determined.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The general and efficient cross-coupling of thiols with aryl halides was developed utilizing Pd(OAc) 2 /1,1′-bis(diisopropylphosphino)ferrocene as the catalyst. The oxidative electrochemistry of dippf was examined, and it was found to follow an ECDim mechanism. Seven new compounds containing dippf were prepared and characterized spectroscopically. The crystal structures of [MCl2(dippf)] (M = Pt and Zn) were determined.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97239-80-0
Molecular Formula C22H36FeP2
Molecular Weight (g/mol) 418.32
MDL Number MFCD02684559
IUPAC Name 1,1'-Bis(diisopropylphosphino)ferrocene
SMILES [Fe].CC(C)P(C(C)C)c1cccc1.CC(C)P(C(C)C)c1cccc1

Specifications

Melting Point 50°C to 52°C
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 418.34
Percent Purity 98%
Chemical Name or Material 1,1'-Bis(diisopropylphosphino)ferrocene
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.