accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OCH3CH3HOOSOHNO

CAS RN 97322-87-7

IUPAC Name: 5-({4-[(6-hydroxy-2,5,7,8-tetramethyl-3,4-dihydro-2H-1-benzopyran-2-yl)methoxy]phenyl}methyl)-1,3-thiazolidine-2,4-dione
Synonyms: troglitazone rezulin (TN) troglitazona romglizone troglitazonum
Molecular Weight (g/mol): 441.54
Molecular Formula: C24H27NO5S
InChi Key: GXPHKUHSUJUWKP-UHFFFAOYNA-N
SMILES: CC1=C(C)C2=C(CCC(C)(COC3=CC=C(CC4SC(=O)NC4=O)C=C3)O2)C(C)=C1O
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results