accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOOHO

CAS RN 97534-82-2

IUPAC Name: 3-[(benzyloxy)carbonyl]-1,3-oxazolidine-4-carboxylic acid
Synonyms: 3-[(benzyloxy)carbonyl]-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 251.24
Molecular Formula: C12H13NO5
InChi Key: XRRRGBIMHQARMF-UHFFFAOYNA-N
SMILES: OC(=O)C1COCN1C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results