accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOHO

CAS RN 97744-77-9

IUPAC Name: 2-{6-oxo-2H,6H-[1,3]dioxolo[4,5-g]chromen-8-yl}acetic acid
Synonyms: {6-oxo-2H-[1,3]dioxolo[4,5-g]chromen-8-yl}acetic acid
Molecular Weight (g/mol): 248.19
Molecular Formula: C12H8O6
InChi Key: PIHRLFGNZWHKIO-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(=O)OC2=CC3=C(OCO3)C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results