CAS RN 98-95-3
Nitrobenzene, 99.5%, for analysis
CAS: 98-95-3 Molecular Formula: C6H5NO2 Molecular Weight (g/mol): 123.11 InChI Key: LQNUZADURLCDLV-UHFFFAOYSA-N Synonym: nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene PubChem CID: 7416 ChEBI: CHEBI:27798 IUPAC Name: nitrobenzene SMILES: C1=CC=C(C=C1)[N+](=O)[O-]
Nitrobenzene, 99%
CAS: 98-95-3 Molecular Formula: C6H5NO2 Molecular Weight (g/mol): 123.111 MDL Number: MFCD00007043 InChI Key: LQNUZADURLCDLV-UHFFFAOYSA-N Synonym: nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene PubChem CID: 7416 ChEBI: CHEBI:27798 IUPAC Name: nitrobenzene SMILES: C1=CC=C(C=C1)[N+](=O)[O-]
Nitrobenzene, 99+%, ACS reagent
CAS: 98-95-3 Molecular Formula: C6H5NO2 Molecular Weight (g/mol): 123.11 MDL Number: MFCD00007043 InChI Key: LQNUZADURLCDLV-UHFFFAOYSA-N Synonym: nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene PubChem CID: 7416 ChEBI: CHEBI:27798 IUPAC Name: nitrobenzene SMILES: C1=CC=C(C=C1)[N+](=O)[O-]
Nitrobenzene, 99%, Extra Pure
CAS: 98-95-3 Molecular Formula: C6H5NO2 Molecular Weight (g/mol): 123.11 MDL Number: MFCD00007043 InChI Key: LQNUZADURLCDLV-UHFFFAOYSA-N Synonym: nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene PubChem CID: 7416 ChEBI: CHEBI:27798 IUPAC Name: nitrobenzene SMILES: C1=CC=C(C=C1)[N+](=O)[O-]
Nitrobenzene, 100.0%, 99.0% purity, For GC analysis, MP Biomedicals™
CAS: 98-95-3 Molecular Formula: C6H5NO2 Molecular Weight (g/mol): 123.11 InChI Key: LQNUZADURLCDLV-UHFFFAOYSA-N Synonym: benzene, nitro,nitro-benzene,unii-zi9w9e8g2z,nob,zi9w9e8g2z,chembl98797,nitroso benzene,keto-aniline,nitrosobenzene,acmc-209m6j PubChem CID: 11473 ChEBI: CHEBI:27986 IUPAC Name: nitrobenzene SMILES: [O-][N+](=O)C1=CC=CC=C1
PESTANAL™ Nitrobenzene, ≥99% (GC), MilliporeSigma™ Supelco™
Nitrobenzene is a carcinogenic compound which has been employed extensively in dyes, pesticides and explosives industries.
Nitrobenzene 99.5+%, TCI America™
CAS: 98-95-3 Molecular Formula: C6H5NO2 Molecular Weight (g/mol): 123.111 InChI Key: LQNUZADURLCDLV-UHFFFAOYSA-N Synonym: nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene PubChem CID: 7416 ChEBI: CHEBI:27798 IUPAC Name: nitrobenzene SMILES: C1=CC=C(C=C1)[N+](=O)[O-]
Nitrobenzene (Certified ACS), Fisher Chemical™
CAS: 98-95-3 Molecular Formula: C6H5NO2 Molecular Weight (g/mol): 123.111 MDL Number: MFCD00007043 InChI Key: LQNUZADURLCDLV-UHFFFAOYSA-N Synonym: nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene PubChem CID: 7416 ChEBI: CHEBI:27798 IUPAC Name: nitrobenzene SMILES: C1=CC=C(C=C1)[N+](=O)[O-]
Nifuroxazide, 99.9%, MP Biomedicals™
CAS: 98-95-3 Molecular Formula: C6H5NO2 Molecular Weight (g/mol): 123.111 InChI Key: LQNUZADURLCDLV-UHFFFAOYSA-N Synonym: nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene PubChem CID: 7416 ChEBI: CHEBI:27798 IUPAC Name: nitrobenzene SMILES: C1=CC=C(C=C1)[N+](=O)[O-]
Nitrobenzene, SPEX CertiPrep™
CAS: 98-95-3 Molecular Formula: C6H5NO2 Molecular Weight (g/mol): 123.111 InChI Key: LQNUZADURLCDLV-UHFFFAOYSA-N PubChem CID: 7416 ChEBI: CHEBI:27798 IUPAC Name: nitrobenzene SMILES: C1=CC=C(C=C1)[N+](=O)[O-]