Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrobenzene, 100.0%, 99.0% purity, For GC analysis, MP Biomedicals™

Catalog No. ICN15585190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15585190 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15585190 Supplier MP Biomedicals, Inc Supplier No. 0215585190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym benzene, nitro, nitro-benzene, unii-zi9w9e8g2z, nob, zi9w9e8g2z, chembl98797, nitroso benzene, keto-aniline, nitrosobenzene, acmc-209m6j
PubChem CID 11473
ChEBI CHEBI:27986
IUPAC Name nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1

Specifications

Color Yellow
Density 1.196g/mL at 25°C
Assay Percent Range 99%
Quantity 500 mL
Formula Weight 123.12
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material Nitrobenzene, For GC analysis
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.