accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNH2HCl

CAS RN 98071-16-0

IUPAC Name: 1-aminocyclobutane-1-carboxylic acid hydrochloride
Synonyms: 1-aminocyclobutane-1-carboxylic acid hydrochloride
Molecular Weight (g/mol): 151.59
Molecular Formula: C5H10ClNO2
InChi Key: HBTVGNDTGRUBQO-UHFFFAOYSA-N
SMILES: Cl.NC1(CCC1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results