CAS RN 98319-26-7
Finasteride
CAS: 98319-26-7 Molecular Formula: C23H36N2O2 Molecular Weight (g/mol): 372.553 MDL Number: MFCD00869737 InChI Key: DBEPLOCGEIEOCV-WSBQPABSSA-N Synonym: finasteride,proscar,propecia,finastid,prostide,chibro-proscar,finasterida,finasteridum,finpecia,finasteridum inn-latin PubChem CID: 57363 ChEBI: CHEBI:5062 IUPAC Name: (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide SMILES: CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C
Finasteride, USP, 98.5-101%, Spectrum™ Chemical
CAS: 98319-26-7 Molecular Formula: C23H36N2O2 Molecular Weight (g/mol): 372.55 InChI Key: DBEPLOCGEIEOCV-WSBQPABSSA-N IUPAC Name: (4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-tert-butyl-4a,6a-dimethyl-2-oxo-1H,2H,4aH,4bH,5H,6H,6aH,7H,8H,9H,9aH,9bH,10H,11H,11aH-indeno[5,4-f]quinoline-7-carboxamide SMILES: CC(C)(C)NC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
Thermo Scientific Chemicals Finasteride, 98%
CAS: 98319-26-7 Molecular Formula: C23H36N2O2 Molecular Weight (g/mol): 372.54 InChI Key: DBEPLOCGEIEOCV-WSBQPABSSA-N Synonym: finasteride,proscar,propecia,finastid,prostide,chibro-proscar,finasterida,finasteridum,finpecia,finasteridum inn-latin PubChem CID: 57363 ChEBI: CHEBI:5062 IUPAC Name: (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide SMILES: CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C
Finasteride 98.0+%, TCI America™
CAS: 98319-26-7 Molecular Formula: C23H36N2O2 Molecular Weight (g/mol): 372.553 MDL Number: MFCD00869737 InChI Key: DBEPLOCGEIEOCV-WSBQPABSSA-N Synonym: finasteride,proscar,propecia,finastid,prostide,chibro-proscar,finasterida,finasteridum,finpecia,finasteridum inn-latin PubChem CID: 57363 ChEBI: CHEBI:5062 IUPAC Name: (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide SMILES: CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C
R&D Systems™ Finasteride
CAS: 98319-26-7 Molecular Formula: C23H36N2O2