Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Finasteride, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18602731 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-731 100 g Amber Glass Bottle
18-602-728 5 g Amber Glass Bottle
18-602-729 25 g Amber Glass Bottle
18-602-730 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-731 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1300100GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

F1300, 98319-26-7, C23H36N2O2

Spectrum™ Chemical Finasteride, USP

Chemical Identifiers

CAS 98319-26-7
Molecular Formula C23H36N2O2
Molecular Weight (g/mol) 372.55
InChI Key DBEPLOCGEIEOCV-WSBQPABSSA-N
IUPAC Name (4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-tert-butyl-4a,6a-dimethyl-2-oxo-1H,2H,4aH,4bH,5H,6H,6aH,7H,8H,9H,9aH,9bH,10H,11H,11aH-indeno[5,4-f]quinoline-7-carboxamide
SMILES CC(C)(C)NC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Melting Range 252°C to 254°C
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation -56.0° to -60.0°
Percent Purity 98.5 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Finasteride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.