CAS RN 98625-26-4
IUPAC Name:
methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
Synonyms:
methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
Molecular Weight (g/mol):
356.3
Molecular Formula:
C16H15F3N2O4
InChi Key:
ZFLWDHHVRRZMEI-UHFFFAOYNA-N
SMILES:
COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1C(F)(F)F)[N+]([O-])=O
1
–
1
of
1
results
1
R&D Systems™ (S)-(-)-Bay K 8644
CAS: 98625-26-4 Molecular Formula: C16H15F3N2O4