accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNH2NNNN(R)O(R)OPOHOOPOOOPHOOO(S)HO(R)HO

CAS RN 987-65-5

IUPAC Name: disodium hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
Synonyms: disodium adenosine triphosphate
Molecular Weight (g/mol): 551.15
Molecular Formula: C10H14N5Na2O13P3
InChi Key: TTWYZDPBDWHJOR-IDIVVRGQSA-L
SMILES: [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)