CAS RN 98769-84-7
IUPAC Name:
(2R)-2-[(R)-(2-ethoxyphenoxy)(phenyl)methyl]morpholine; methanesulfonic acid
Synonyms:
methanesulfonic acid; reboxetine
Molecular Weight (g/mol):
409.5
Molecular Formula:
C20H27NO6S
InChi Key:
CGTZMJIMMUNLQD-STYNFMPRSA-N
SMILES:
CS(O)(=O)=O.CCOC1=CC=CC=C1O[C@@H]([C@H]1CNCCO1)C1=CC=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ Reboxetine mesylate
CAS: 98769-84-7 Molecular Formula: C20H27NO6S