accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99-65-0

IUPAC Name: 1,3-dinitrobenzene
Synonyms: 2,4-dinitrobenzene m-dnb dinitrophenylene
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: WDCYWAQPCXBPJA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)