Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dinitrobenzene 98%, Thermo Scientific™

Catalog No. AC116971000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC116971000 100 g
AC116970250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC116971000 Supplier Thermo Scientific Chemicals Supplier No. 116971000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-65-0
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
InChI Key WDCYWAQPCXBPJA-UHFFFAOYSA-N
Synonym m-dinitrobenzene, 2,4-dinitrobenzene, benzene, 1,3-dinitro, meta-dinitrobenzene, 1,3-dinitrobenzol, dinitrobenzene, benzene, m-dinitro, dwunitrobenzen, m-dnb, dinitrophenylene
PubChem CID 7452
ChEBI CHEBI:51397
IUPAC Name 1,3-dinitrobenzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 86°C to 90°C
Color Brown to Brown-Orange
Density 1.575g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula C6H4(NO2)2
Quantity 100 g
Beilstein 05, 258
Merck Index 15, 3299
Formula Weight 168.11
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 1, 3-Dinitrobenzene, 98%
Show More Show Less
EINECSNumber 202-776-8
RTECSNumber CZ7350000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.