accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99281-50-2

IUPAC Name: (1S,2R)-N1,N1-bis(butan-2-yl)-N2,N2-dicyclohexylcyclohexane-1,2-dicarboxamide
Synonyms: (1R,2S)-N1,N1-dicyclohexyl-N2,N2-bis(sec-butyl)cyclohexane-1,2-dicarboxamide
Molecular Weight (g/mol): 446.72
Molecular Formula: C28H50N2O2
InChi Key: HEMVYQNWBJZPTO-BDBAHPOVSA-N
SMILES: CCC(C)N(C(C)CC)C(=O)[C@H]1CCCC[C@H]1C(=O)N(C1CCCCC1)C1CCCCC1
Grade 
  • (1)
Product Line 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results