accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 993-12-4

IUPAC Name: chlorodimethyl-λ⁵-phosphanethione
Synonyms: chlorodimethyl-λ⁵-phosphanethione
Molecular Weight (g/mol): 128.55
Molecular Formula: C2H6ClPS
InChi Key: UGOLAPHJCTVIEW-UHFFFAOYSA-N
SMILES: CP(C)(Cl)=S
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results