accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 99584-10-8

IUPAC Name: 6-amino-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 6-amino-3-methyl-1,3-benzoxazol-2-one
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: FPNLXQSOWBNXCN-UHFFFAOYSA-N
SMILES: CN1C(=O)OC2=CC(N)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results