CAS RN 99685-96-8
Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2%
CAS: 99685-96-8 Molecular Formula: C60 Molecular Weight (g/mol): 720.66 MDL Number: MFCD00282904 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Synonym: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, 99.5% C{60}
CAS: 99685-96-8 Molecular Formula: C60 Molecular Weight (g/mol): 720.66 MDL Number: MFCD00151408 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Synonym: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene C60, 99.9%
CAS: 99685-96-8 Molecular Formula: C60 Molecular Weight (g/mol): 720.66 MDL Number: MFCD00151408 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Synonym: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, hydroxylated, C{60}(OH)∣n,
CAS: 99685-96-8 Molecular Formula: C60 Molecular Weight (g/mol): 720.66 MDL Number: MFCD06202029 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Synonym: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, 99.9+% C{60}
CAS: 99685-96-8 Molecular Formula: C60 Molecular Weight (g/mol): 720.66 MDL Number: MFCD00282904 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Synonym: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, sublimed, 99.9+% C{60}
CAS: 99685-96-8 Molecular Formula: C60 Molecular Weight (g/mol): 720.66 MDL Number: MFCD00151408 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Synonym: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene powder, 99% C{6}{0}
CAS: 99685-96-8 Molecular Formula: C60 Molecular Weight (g/mol): 720.66 MDL Number: MFCD00282904 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Synonym: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C\{60}-I\{h})[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene C60 (pure) 99.5+%, TCI America™
CAS: 99685-96-8 Molecular Formula: C60 Molecular Weight (g/mol): 720.66 MDL Number: MFCD00151408 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Synonym: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C60-Ih)[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Fullerene C60 99.0+%, TCI America™
CAS: 99685-96-8 Molecular Formula: C60 Molecular Weight (g/mol): 720.66 MDL Number: MFCD00151408 InChI Key: XMWRBQBLMFGWIX-UHFFFAOYSA-N Synonym: fullerene,buckminsterfullerene,buckyball,fullerene 60,60 fullerene,footballene,fullerene-c60,5,6 fullerene-c60-ih PubChem CID: 123591 ChEBI: CHEBI:33128 IUPAC Name: (C60-Ih)[5,6]fullerene SMILES: C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Sigma Aldrich Fullerene-C60
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Fullerene-C60
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Fullerene-C60
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.