accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2OOOOCH2(Z)

CAS RN 999-21-3

IUPAC Name: 1,4-bis(prop-2-en-1-yl) (2Z)-but-2-enedioate
Synonyms: diallyl maleate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: ZPOLOEWJWXZUSP-WAYWQWQTSA-N
SMILES: C=CCOC(=O)\C=C/C(=O)OCC=C
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)