Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diallyl maleate, 95%

Catalog No. AAL0386730
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0386730 250 g
AAL038670B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0386730 Supplier Thermo Scientific Chemicals Supplier No. L0386730
  • $160.40
    $131.65 / Each of 1

    Save $28.75

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

CAS: 999-21-3 | C10H12O4 | 196.202 g/mol

Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.

Solubility
Soluble in water 151.2 mg/L @ 25°C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-21-3
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00010763
InChI Key ZPOLOEWJWXZUSP-WAYWQWQTSA-N
Synonym diallyl fumarate, fumaric acid, diallyl ester, unii-k852lk4c5p, 2-butenedioic acid e-, di-2-propenyl ester, 2-butenedioic acid 2e-, di-2-propenyl ester, butenedioic acid diallyl ester, 1,4-bis prop-2-en-1-yl 2e-but-2-enedioate, 2-butenedioic acid 2e-, 1,4-di-2-propen-1-yl ester, diallyl malleate, 1,4-bis prop-2-en-1-yl but-2-enedioate
PubChem CID 5354266
IUPAC Name bis(prop-2-enyl) (Z)-but-2-enedioate
SMILES C=CCOC(=O)C=CC(=O)OCC=C

Specifications

Melting Point -47°C
Density 1.072
Boiling Point 109°C to 110°C (3 mmHg)
Flash Point 130°C (266°F)
Refractive Index 1.467
Quantity 250 g
UN Number UN2810
Beilstein 1725954
Solubility Information Soluble in water 151.2mg/L @ 25°C.
Formula Weight 196.21
Percent Purity 95%
Chemical Name or Material Diallyl maleate
Show More Show Less
Hazard Category H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 213-658-0
RTECSNumber ON0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.