UserName
Viewing profile as user:
Applied Biosystems SeqStudio Genetic Analyzer
Applied Biosystems Ion GeneStudio S5 System
Applied Biosystems SeqStudio Genetic Analyzer
Save Time in Your Workflow
Trust Applied Biosystems Instruments to Meet Your Needs
Applied Biosystems™ products offer an affordable yet modern experience, providing you with what you need to enhance your lab productivity. Use the Applied Biosystems™ SeqStudio™ Genetic Analyzer for a wide range of sequencing and fragment analysis applications to deliver high-quality results in minutes instead of hours.
Innovative
Just click. That's it
The simple design of the SeqStudio Genetic Analyzer makes setup and maintenance easy. No need to calibrate before each run - just load your samples, click the cartridge in place and begin. The all-in-one cartridge uses a single polymer for both sequencing and fragment analysis applications and is stable on the instrument for up to 4 months.
Flexible
Gain flexibility in your lab by performing both Sanger sequencing and fragment analysis on the same plate with the SeqStudio Genetic Analyzer.
Fast
Quick sample turnaround with run times in as little as 30 minutes.
Easy to Use and Share
The SeqStudio Genetic Analyzer uses an all-in-one cartridge to give researchers flexibility when designing experiments. The cartridge is convenient and efficient and allows you to use only what you need and avoid unnecessary waste. The unique cartridge design enables multiple users to share a single SeqStudio Genetic Analyzer. The instrument has an interactive touch screen that makes setup and data viewing easy. Training is also included with purchase.
Go Mobile
Access your data with ease using web-based software from the Thermo Fisher Cloud.  
  • Set up and monitor your run, then manage your data from any location using your phone, tablet, or computer
  • Use the Instrument Connect mobile app to check progress, monitor your results, and perform primary and secondary analysis
  • Share data with colleagues using secure connections 
For Research Use Only. Not for use in diagnostic procedures.
Contact an Account Manager
Complete this form to contact a Fisher Healthcare representative about the Applied Biosystems SeqStudio Genetic Analyzer.
Required Field*
State / Province
Alaska
Alabama
Arkansas
Arizona
California
Colorado
Connecticut
D.C.
Florida
Georgia
Guam
Hawaii
Iowa
Idaho
Illinois
Indiana
Kansas
Kentucky
Louisiana
Massachusetts
Maryland
Maine
Michigan
Minnesota
Missouri
Mississippi
Montana
North Carolina
North Dakota
Nebraska
New Hampshire
New Jersey
New Mexico
Nevada
New York
Ohio
Oklahoma
Oregon
Pennsylvania
Puerto Rico
Palau
Rhode Island
South Carolina
South Dakota
Tennessee
Texas
Utah
Virginia
Virgin Islands
Vermont
Washington
Wisconsin
West Virginia
Wyoming
Military Americas
Military Europe/ME/Canada
Military Pacific
Alberta
Manitoba
British Columbia
New Brunswick
Newfoundland and Labrador
Nova Scotia
Northwest Territories
Nunavut
Ontario
Prince Edward Island
Quebec
Saskatchewan
Yukon Territory
By submitting your data, you agree to our Privacy Policy and will receive emails based on your preferences.
© Thermo Fisher Scientific, Inc. All Rights Reserved.
Thermo Fisher Scientific Logo
Save Time In Your Workflow
Figure 1. SeqStudio Genetic Analyzer Workflow