UserName
Viewing profile as user:
Applied Biosystems Ion GeneStudio S5 System
Applied Biosystems Ion GeneStudio S5 System
Ion GeneStudio S5 System
Reduce Setup Time and Complexity
Set up simple, targeted workflows for assessing microbial genomes, gene panels, exomes, and transcriptomes with the Ion GeneStudio™ S5 next-generation sequencing system. Its cartridge-based reagents and Ion 5 Series Chips make it easy to use and support a range of high-throughput sequencing for clinical research and other applications. Plus, the GeneStudio S5 brings flexible, affordable sequencing to labs that work with smaller panels and have lower sequencing throughput requirements.
  • Ion 5 Series Chips can achieve from 2 to 80 million reads per run
  • Cartridge-based, plug-and-play reagents help to reduce setup time
  • Run and analysis complete in hours
Contact an Account Manager
Fill out this form to get more information about Applied Biosystems and Ion Torrent qPCR, CE, and NGS products.
Required Field*
State / Province
Alaska
Alabama
Arkansas
Arizona
California
Colorado
Connecticut
D.C.
Florida
Georgia
Guam
Hawaii
Iowa
Idaho
Illinois
Indiana
Kansas
Kentucky
Louisiana
Massachusetts
Maryland
Maine
Michigan
Minnesota
Missouri
Mississippi
Montana
North Carolina
North Dakota
Nebraska
New Hampshire
New Jersey
New Mexico
Nevada
New York
Ohio
Oklahoma
Oregon
Pennsylvania
Puerto Rico
Palau
Rhode Island
South Carolina
South Dakota
Tennessee
Texas
Utah
Virginia
Virgin Islands
Vermont
Washington
Wisconsin
West Virginia
Wyoming
Military Americas
Military Europe/ME/Canada
Military Pacific
Alberta
Manitoba
British Columbia
New Brunswick
Newfoundland and Labrador
Nova Scotia
Northwest Territories
Nunavut
Ontario
Prince Edward Island
Quebec
Saskatchewan
Yukon Territory
By submitting your data, you agree to our Privacy Policy and will receive emails based on your preferences.
© Thermo Fisher Scientific, Inc. All Rights Reserved.
Thermo Fisher Scientific Logo
The Ion GeneStudio S5 Series
The GeneStudio series includes three options: the S5, the S5 Plus, and the S5 Prime system. They share the same technology and fluidics, but offer different analysis capabilities.
Speed
Ion GeneStudio S5 Sequencers use the power of semiconductors to produce high-quality sequencing data in just a few hours. When paired with the Ion Chef™ System, it can take as little as 24 hours to go from DNA library to data, with only 45 minutes of actual hands-on time.
Simplicity
S5 Sequencers are easy to use, offering single-day installation, a straightforward user interface, and cartridge-based reagents. Combine them with Ion AmpliSeq™ technology (for target selection), the Ion Chef System (for automated library and template preparation), and Ion Reporter™ Software (for automated variant annotation) to make targeted sequencing simpler than ever.
Small Sample Input
Use as little as 1ng of DNA or RNA from a variety of sample types, including formalin-fixed paraffin-embedded (FFPE) tissue, retrospective samples from fine needle aspirates, and cell-free DNA extracted from blood (cfDNA).
Scalability
All of your targeted sequencing or microbial applications can be performed on a single GeneStudio S5 Sequencer using different Ion 5 Series Chips. This flexibility provides several throughput options to let you transition from small to large projects.
Analysis and Storage Solutions
Make NGS even easier with Torrent Suite and Ion Reporter Software. Plan, monitor, track, and analyze your runs using Torrent Suite Software (pre-installed on the Torrent Server). Then integrate, annotate, and interpret variants using Ion Reporter Software (used locally or from the Thermo Fisher Cloud).
Services and Support
With an extensive network of more than 2,000 dedicated field application specialists and service engineers, we’ll be there when and where you need us.
  • Service and support plans — maximize instrument performance and ensure availability of critical systems with comprehensive plans
  • Qualification services — document and ensure your Ion GeneStudio S5 is installed, operating, and performing to the manufacturer’s specifications
  • Training — learn about experimental design, data analysis, troubleshooting, applications, and instrument operation
  • Bioinformatics — get tailored support for software, data analysis, execution, and downstream applications
  • DataSafe solution — implement storage design and backup
FAQ
Q: Can the GeneStudio S5 run two chips at the same time?
A: No. Analysis with the first chip must be complete before sequencing with a second chip can begin.
Q: Does the GeneStudio S5 have a separate analysis server?
A: No. The analysis server is contained within the sequencer itself.
Q: How long does it take to run an Ion 540 Chip on a GeneStudio S5?
A: 19 hours.
Q: Is the GeneStudio S5 compatible with the Ion Chef System?
A: Yes.
Flexible Throughput and Workflows
Chip Type
Number of Reads
Read Length (Output)
Turnaround Time (Sequencing Run and Analysis Time)
Ion GeneStudio S5 System
Ion GeneStudio S5 Plus System
Ion GeneStudio S5 Prime System
Ion 510 Chip
2–3M
200 bp (0.3–0.5 Gb)
4.5 hr
3 hr
3 hr
400 bp (0.6–1 Gb)
10.5 hr
5 hr
5 hr
Ion 520 Chip
4–6M
200 bp (0.6–1 Gb)
7.5 hr
3.5 hr
3 hr
400 bp (1.2–2 Gb)
12 hr
5.5 hr
5.5 hr
3–4M
600 bp (0.5–1.5 Gb)
12 hr
5.5 hr
5.5 hr
Ion 530 Chip
15–20M
200 bp (3–4 Gb)
10.5 hr
5 hr
4 hr
400 bp (6–8 Gb)
21.5 hr
8 hr
6.5 hr
9–12M
600 bp (1.5–4.5 Gb)
21 hr
8 hr
7 hr
Ion 540 Chip
60–80M
200 bp (10–15 Gb)
19 hr
10 hr
6.5 hr
200 bp (20–30 Gb), 2 runs in 1 day
N/A
20 hr
10 hr
Ion 550 Chip
100–130M
200 bp (20–25 Gb)
N/A
11.5 hr
8.5 hr
200 bp (40–50 Gb), 2 runs in 1 day
N/A
N/A
12 hr
For research use only. Not for use in diagnostic procedures.