Fisherbrand Traceable Datalogging Devices
Fisherbrand™ Traceable™ Min/Max Thermometers
- Min/max memory
- Great for all applications
Key benefits:
- Event storage
- Audible/visual alarm/notification when limits have been crossed
Applications/usage:
Min/max thermometers specifically measure the minimum and maximum temperature to which the thermometer is exposed. This information is used to determine exposure of the product to temperature extremes or environmental temperature fluctuations.
Learn More
Datalogger Thermometers
Fisherbrand™ Traceable™ Excursion-Trac™ Datalogger Thermometers- Erasable memory; USB data downloads
- Rolling memory stores > 526,000 data points
- Permanent data memory; USB data downloads
- 21 CFR 11 environments
Key benefits:
- Transfer data in CSV format via USB drive —no software required
- Audible/visual alarms when parameters have been breached
- Meets CDC/VFC requirements
Applications/usage:
For sensitive environments such as refrigerator/freezers, ultra-low-temperature freezers, vaccines, chemical storerooms, biofuel research, greenhouses, liquid nitrogen storage, microbiology/immunology, blood banks, food and beverage, and controlled warehouses.
Learn More
Fisherbrand™ TraceableLIVE™ Datalogger Thermometers
- 24/7 worry-free monitoring via WIFI
- Real-time email, text or push notifications of parameter breaches
Key benefits:
- View data from anywhere; run reports via app or internet
- Unlimited scalability and number of users
- Meets CDC/VFC requirements
Applications/usage:
Use in critical environments such as refrigerator/freezers, ultra-low-temperature freezers, vaccines, chemical storerooms, biofuel research, greenhouses, liquid nitrogen storage, microbiology/immunology, blood banks, food and beverage, and controlled warehouses.
Learn MoreFisherbrand Traceable internal memory datalogging devices allow you to get a complete and continuous history of your measurements without cumbersome and unreliable manual record keeping.
- When temperatures are out of range, the Smart-Alarm™ feature alerts you visually and audibly (until you turn it off)
- Units with the MemoryLoc™ feature can store more than 1 million data points (about two years of data) and will stop recording when the memory is full (no data overwriting)
- Transfer recorded data (CSV file) using a USB flash drive (not included)
- Units with the Excursion-Trac™ feature maintain the most recent 525,600 temperature observations (rolling memory)
Fisherbrand Traceable Dataloggers
Find out how Traceable digital dataloggers allow you to stop manually recording data and avoid errors.
Datalogging
Put Down the Pen
For each thermometer, reports show the exact temperature readings of probe(s) with time and day stamp, as well as the exact time and date when the units go outside of user-defined set points and when the units come back into range.
Analyze and Trend Monitoring Data
Take your observations and turn them into meaningful information. Access to recorded data allows you to determine how temperature readings are trending and gives you the ability to analyze and form conclusions about storage and laboratory conditions.