Fisherbrand Traceable Fundamental Monitoring Devices
Fisherbrand™ Traceable™ Min/Max Thermometers
- Min/max memory
- Great for all applications
Key benefits:
- Event storage
- Audible/visual alarm/notification when limits have been crossed
Applications/usage:
Min/max thermometers specifically measure the minimum and maximum temperature to which the thermometer is exposed. This information is used to determine exposure of the product to temperature extremes or environmental temperature fluctuations.
Learn More
Datalogger Thermometers
Fisherbrand™ Traceable™ Excursion-Trac™ Datalogger Thermometers- Erasable memory; USB data downloads
- Rolling memory stores > 526,000 data points
- Permanent data memory; USB data downloads
- 21 CFR 11 environments
Key benefits:
- Transfer data in CSV format via USB drive —no software required
- Audible/visual alarms when parameters have been breached
- Meets CDC/VFC requirements
Applications/usage:
For sensitive environments such as refrigerator/freezers, ultra-low-temperature freezers, vaccines, chemical storerooms, biofuel research, greenhouses, liquid nitrogen storage, microbiology/immunology, blood banks, food and beverage, and controlled warehouses.
Learn More
Fisherbrand™ TraceableLIVE™ Datalogger Thermometers
- 24/7 worry-free monitoring via WIFI
- Real-time email, text or push notifications of parameter breaches
Key benefits:
- View data from anywhere; run reports via app or internet
- Unlimited scalability and number of users
- Meets CDC/VFC requirements
Applications/usage:
Use in critical environments such as refrigerator/freezers, ultra-low-temperature freezers, vaccines, chemical storerooms, biofuel research, greenhouses, liquid nitrogen storage, microbiology/immunology, blood banks, food and beverage, and controlled warehouses.
Learn MoreFisherbrand Traceable monitoring devices always come with an individually serialized multi-point Traceable Calibration Certificate from an ISO/IEC 17025:2005 (1750.01) calibration laboratory indicating traceability to NIST standards.
Features include:
- Min/max memory
- Programmable high/low alarm settings
- Audible and visual alarms
- Accuracies ranging from ±1° to ±0.05°C
Choose from a variety of Traceable thermometers that are suitable for use with liquids, gases, and semi-solids and allow you to obtain accurate records for most temperature conditions and environments.
Traceable thermometers offer minimum and maximum memories and feature visual LED or audible alarms. Many of the units also allow you to program high or low alarm settings. So whether you are interested in simply monitoring laboratory conditions or taking care of your precious samples in refrigerators or freezers, we have the perfect solution to meet your needs.