Fisherbrand Traceable Thermometers
Explore Devices and Features
- Fundamental Monitoring Devices
- Datalogging Devices
- Lab Monitoring Devices
Fisherbrand Traceable monitoring devices always come with an individually serialized multi-point Traceable Calibration Certificate from an ISO/IEC 17025:2005 (1750.01) calibration laboratory indicating traceability to NIST standards.
Features include:
- Min/max memory
- Programmable high/low alarm settings
- Audible and visual alarms
- Accuracies ranging from ±1° to ±0.05°C
Choose from a variety of Traceable thermometers that are suitable for use with liquids, gases, and semi-solids and allow you to obtain accurate records for most temperature conditions and environments.
Traceable thermometers offer minimum and maximum memories and feature visual LED or audible alarms. Many of the units also allow you to program high or low alarm settings. So whether you are interested in simply monitoring laboratory conditions or taking care of your precious samples in refrigerators or freezers, we have the perfect solution to meet your needs.
Featured Products
Catalog Number 0666411
Catalog Number 14650103
Catalog Number 15081109
Catalog Number 15081102
Fisherbrand Traceable internal memory datalogging devices allow you to get a complete and continuous history of your measurements without cumbersome and unreliable manual record keeping.
- When temperatures are out of range, the Smart-Alarm™ feature alerts you visually and audibly (until you turn it off)
- Units with the MemoryLoc™ feature can store more than 1 million data points (about two years of data) and will stop recording when the memory is full (no data overwriting)
- Transfer recorded data (CSV file) using a USB flash drive (not included)
- Units with the Excursion-Trac™ feature maintain the most recent 525,600 temperature observations (rolling memory)
Fisherbrand Traceable Dataloggers
Find out how Traceable digital dataloggers allow you to stop manually recording data and avoid errors.
Datalogging
Put Down the Pen
For each thermometer, reports show the exact temperature readings of probe(s) with time and day stamp, as well as the exact time and date when the units go outside of user-defined set points and when the units come back into range.
Analyze and Trend Monitoring Data
Take your observations and turn them into meaningful information. Access to recorded data allows you to determine how temperature readings are trending and gives you the ability to analyze and form conclusions about storage and laboratory conditions.
Featured Products
Catalog Number 15081123
Catalog Number 15081130
Live and Secure Remote Monitoring Wherever You Go
Setting up TraceableLIVE services couldn't be easier and doesn't require any additional software or hardware. Simply purchase a TraceableLIVE product, connect it to your existing Wi-Fi network, set up an account through the TraceableLIVE website on any browser, and point your device to your account. Within a few minutes, your device will begin to transmit critical data to your personal cloud, and you can also control your device through your smartphone, tablet, or computer.
The scalability and segmentability of TraceableLIVE knows no end. Add as many devices as you want and divide them into as many groups as you want.
Fisherbrand TraceableLIVE Monitoring Services
See how TraceableLIVE Monitoring Services allow you to stay connected to your critical environments, wherever you go.
Wirelessly Monitor Temperature, Humidity, CO2 and Barometric Pressure
With Fisherbrand TraceableLIVE, you’re always connected to your cold storage or ambient environments through your smartphone, tablet, or personal computer. Simply use the TraceableLIVE app or visit TraceableLIVE.com to:
- View current conditions in your laboratory at any time
- Control your alarm parameters
- View your datalogging history
- Generate and email reports
- Give other team members access to groups of devices or specific devices
- And so much more
Instant Notification
With mobile push, email, and text functionality, TraceableLIVE ensures peace of mind by protecting your work. If any of your TraceableLIVE devices exceed an alarm parameter, lose connection to your Wi-Fi signal, or begin to run low on battery life, TraceableLIVE will immediately send you alerts based on your indicated preferences, which may include:
- Mobile push
- Text message
Event Documentation
A permanent record with detailed information will be created if a Fisherbrand TraceableLIVE device experiences:
- An alarm event
- A change in alarm parameters
- A low battery alert
- A loss in Wi-Fi connectivity
Change Control
With documented Change Control, you'll have permanent records of the events that have occurred to your Fisherbrand TraceableLIVE devices and your environments.
Unlimited Scalability and Segmentability
Add as many devices to your TraceableLIVE account as you want and divide them into as many groups as you want. Then, using your administrative controls, provide specific individuals access to certain devices or groups of devices. This way, only the data that’s relevant to others will appear on their own accounts, while you continue to view the status of all the devices at once.
Security and Peace of Mind
Your work is valuable. Protect and preserve your samples and data from accidents, natural disasters, and the unexpected. TraceableLIVE is a cloud-based service using an SSL-encrypted, secure connection to transfer and store your data.
Featured Products
Catalog Number 6960074
Catalog Number 7111647