UserName
Viewing profile as user:
Real-Time PCR Solutions
Real-Time PCR Solutions
Real-time PCR, also known as quantitative or qPCR, is the gold-standard for sensitive, specific detection and quantification of nucleic acid targets.
Applied Biosystems™ qPCR solutions help maximize your time and effort. With well-designed algorithms, optimized master mixes, intuitive data analysis software, and smart instrumentation, we can help you harness the power of real-time PCR.
Applied Biosystems products have been designed with efficiency in mind and work together synergistically to offer you simpler, more flexible, and efficient real-time PCR assays.
Applied Biosystems™ QuantStudio™ Real-Time PCR Systems
Designed to be compatible with Applied Biosystems™ TaqMan™ Assays and with the flexibility of multiple real-time chemistry choices, it’s easy to get your experiments up and running.
QuantStudio instruments let you connect and collaborate with colleagues and get the results you need to achieve your research goals. These real-time PCR platforms offer real value — performance, reliability, and expert technical support.
Visit fishersci.com/quantstudio to learn more about QuantStudio instruments.
Applied Biosystems™ TaqMan™ Products
TaqMan master mixes include buffer, dNTPs, passive reference dye, thermostable hot-start DNA polymerase, and other components formulated for reliable real-time PCR assays. Just add your sample, a TaqMan assay, and the reaction starts.
Visit fishersci.com/qpcrreagents to learn more about TaqMan real-time PCR products.
TaqMan Gene Expression Assays
  • Gold-standard specificity, sensitivity, and reproducibility
  • Faster results with proven TaqMan qPCR chemistry
  • Detect a high number of gene transcripts and transcript variants
  • Access more than 2.8 million predesigned assays
Visit fishersci.com/taqmanassays to shop for Taqman assays.
SYBR Green Products
Applied Biosystems™ SYBR™ Green Master Mixes are designed for quantitative real-time PCR and include a set of two PCR primers that flank the target region. The master mixes contain buffer, dNTPs, thermostable hot-start DNA polymerase, and, of course, SYBR Green dye. Simply provide the sample and PCR primer pair and you'll have everything you need for real-time PCR.
Visit fishersci.com/qpcrreagents to learn more about SYBR Green Master Mix.
Applied Biosystems PowerTrack SYBR Green Master Mix Overview
Discover a qPCR master mix that brings specificity, sensitivity, and efficiency to your research with a two-color tracking dye system.