Filtered Search Results
Gibco™ RPMI 1640 Medium (ATCC modification)
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Gibco™ ACK Lysing Buffer
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Gibco™ StemPro™ Accutase™ Cell Dissociation Reagent
StemPro Accutase is a gentle cell dissociation solution for routine detachment of cells from tissue culture plasticware or adhesion coated plasticware.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Gibco™ Expi293™ Expression Medium
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Expi293 Expression Medium designed for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Product Type | Expression Medium |
|---|---|
| Form | Liquid |
| Classification | Animal Origin-free,Chemically-defined,Protein-free,Serum-free |
| Product Line | Expi293 |
Gibco™ Poly-D-Lysine
Poly-D-Lysine is an extracellular matrix used to facilitate neural cell adhesion to tissue culture-treated plastic and glass surfaces. When used on Nunc plastics with B-27 supplements or the B-27 Plus Neuronal Culture System, Poly-D-Lysine maintains cell viability and normal growth characteristics in multiple neuronal cell types.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Gibco™ Fetal Bovine Serum, Value
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Purity or Quality Grade | Value |
|---|---|
| Sterilization Method | Triple-filtered, 0.1 μm |
| Product Type | Fetal Bovine Serum |
| For Use With (Application) | *FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support. |
| Sterility | Sterile |
| Country of Origin | Refer to CoA for specific origin* |
| Age | Fetal |
Gibco™ Fetal Bovine Serum, Premium
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Purity or Quality Grade | Premium |
|---|---|
| Sterilization Method | Triple-filtered, 0.1 μm |
| Product Type | Fetal Bovine Serum |
| Form | Frozen |
| For Use With (Application) | *FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support. |
| Country of Origin | Refer to CoA for specific origin* |
| Age | Fetal |
Gibco™ ExpiCHO™ Expression Medium
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
A GMP-manufactured, chemically-defined, serum-free, protein-free, animal origin–free medium specifically designed for high-density growth and transfection of suspension-adapted Chinese hamster ovary (CHO) cells.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Shipping Condition | Room Temperature |
|---|---|
| Cell Line | ExpiCHO-S™, CHO-S, SFM Adapted |
| Content And Storage | Storage conditions: in the dark at 2-8°C Shipping conditions: ambient Shelf life: 12 months from date of manufacture |
| With Additives | GlutaMAX |
| Product Type | CHO Expression Medium |
| Form | Liquid |
| Without Additives | No Phenol Red |
| Classification | Animal Origin-free,Chemically-defined,Protein-free,Serum-free |
| Culture Type | Suspension Cell Culture |
| Product Line | ExpiCHO |
| Serum Level | Serum-free |
Gibco™ ExpiFectamine™ CHO Transfection Kit
The ExpiFectamine CHO Transfection Kit is a core component of the ExpiCHO Expression System, designed for transient transfection of high-density cultures of Chinese hamster ovary (CHO) cells.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Shipping Condition | Room Temperature |
|---|---|
| Promoter | pcDNA™ 3.4 |
| Cell Line | ExpiCHO-S™, CHO-S, SFM Adapted |
| Format | Kit |
| Product Type | CHO Transfection Kit |
| Transfection Technique | Lipid-based Transfection |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Product Line | ExpiFectamine |
| Cell Type | Ovarian Cells |
Gibco™ Puromycin Dihydrochloride
Aminonucleoside antibiotic produced by Streptomyces alboniger
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Shipping Condition | Dry Ice |
|---|---|
| Content And Storage | Storage conditions: -5 to -20°C Shipping conditions: Frozen Shelf life: 12 months from date of manufacture |
| With Additives | HEPES |
| Product Type | Antibiotic |
| Form | Liquid |
| Culture Type | Mammalian Cell Culture, Insect Cell Culture |
| Shelf Life | 12 Months |
| Concentration | 10 mg/mL |
| For Use With (Application) | Eukaryotic Selection⁄Stable Cell Line Generation |
| Sterility | Sterile-filtered |
| Product Line | Gibco |
Gibco™ ExpiFectamine™ 293 Transfection Kit
Designed for transient transfection of high density cultures of HEK (human embryonic kidney) 293 cells.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Cell Line | Expi293F™ |
|---|---|
| Product Type | Transfection Kit |
| Transfection Technique | Lipid-based Transfection |
| Sample Type | Plasmid DNA |
| Classification | Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free |
| Serum Compatible | No |
| Purity | Cell Culture Grade |
| For Use With (Application) | Transfection |
| Product Line | ExpiFectamine |
| Cell Type | Established Cell Lines |
Gibco™ Glucose Solution
Gibco™ Glucose Solution contains D-glucose at a concentration of 200 g/L and is used as a serum-free medium supplement suitable for cell culture.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Gibco™ CTS™ DPBS, without calcium chloride, without magnesium chloride
Gibco Cell Therapy Systems (CTS) DPBS is the classic Dulbecco's phosphate-buffered saline. This particular formulation lacks calcium, magnesium, and phenol red.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Includes | No Calcium, No Magnesium |
|---|---|
| Physical Form | Liquid |
| Manufacturing Quality | ISO13485, MDSAP, FDA-registered, 21 CFR820 |
| Chemical Name or Material | DPBS |
| Recommended Storage | Store at 15–30°C. |
| Concentration | 1X |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
| Sterility | Sterile-filtered |
| Solution Type | Dulbecco's Phosphate Buffered Saline |
Gibco™ Fetal Bovine Serum, Premium Plus
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Premium Plus FBS, our most defined FBS, provides consistent cell growth for sensitive cell lines requiring the lowest viral risk.
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
| Sterilization Method | Triple-filtered, 0.1 μm |
|---|---|
| Product Type | Fetal Bovine Serum |
| Form | Frozen |
| For Use With (Application) | *FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support. |
| Sterility | Sterile |
| Country of Origin | Refer to CoA for specific origin* |
| Age | Fetal |
Gibco™ B-27™ Plus Supplement (50X)
B-27 Plus Supplement is the next-generation of the most cited neuronal cell culture product, B-27 Supplement
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More