Assay Details
Target Gene Details
Entrez Gene ID: | 1761 |
Gene Name: | doublesex and mab-3 related transcription factor 1 |
Gene Aliases: |
CT154, DMT1 |
Location: |
Chr.9:841647-969090 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMRT1 | NM_021951.2 | 2 | 620 | NP_068770.2 |
| XM_006716732.1 | 2 | 663 | XP_006716795.1 | |
| XM_011517770.1 | 2 | 556 | XP_011516072.1 | |
| XM_011517771.1 | 2 | 556 | XP_011516073.1 | |
| XM_011517772.1 | 2 | 556 | XP_011516074.1 | |
| XM_011517773.2 | 2 | 375 | XP_011516075.1 | |
| XM_017014374.1 | 2 | 556 | XP_016869863.1 | |
| XM_017014375.1 | 2 | 663 | XP_016869864.1 | |
| AF130728.1 | 2 | 620 | AAD40474.1 | |
| AJ276801.1 | 2 | 529 | CAB82851.1 | |
| AK310336.1 | 2 | 358 | ||
| AK313594.1 | 2 | 613 | ||
| AL162131.1 | 2 | 529 | CAB82427.1 | |
| AY442914.1 | 2 | 620 | AAR89619.1 | |
| BC040847.1 | 2 | 507 | AAH40847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3619193 | Chr.9:514830 - 877545 on Build GRCh38 | Gain |
|
| nsv1023836 | Chr.9:646024 - 877740 on Build GRCh38 | Loss |
|
| nsv831491 | Chr.9:794113 - 974142 on Build GRCh38 | Gain |
|
| nsv613047 | Chr.9:802801 - 855091 on Build GRCh38 | Gain |
|
| esv2759655 | Chr.9:524413 - 853033 on Build GRCh38 | Loss |
|
| nsv1021740 | Chr.9:596826 - 923709 on Build GRCh38 | Gain |
|
| nsv613037 | Chr.9:643453 - 1138368 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map