Assay Details
Target Gene Details
Entrez Gene ID: | 4983 |
Gene Name: | oligophrenin 1 |
Gene Aliases: |
ARHGAP41, MRX60, OPN1 |
Location: |
Chr.X:68042344-68433841 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPHN1 | NM_002547.2 | 25 | 3588 | NP_002538.1 |
| XM_005262270.1 | 24 | 3334 | XP_005262327.1 | |
| XM_011530961.1 | 25 | 3509 | XP_011529263.1 | |
| AJ001189.1 | 25 | 3932 | CAA04579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758871 | Chr.X:67279827 - 68099252 on Build GRCh38 | Gain |
|
| nsv818030 | Chr.X:67496364 - 68089458 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map