Assay Details
Target Gene Details
Entrez Gene ID: | 224 |
Gene Name: | aldehyde dehydrogenase 3 family member A2 |
Gene Aliases: |
ALDH10, FALDH, SLS |
Location: |
Chr.17:19648136-19677596 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH3A2 | NM_000382.2 | NP_000373.1 | ||
| NM_001031806.1 | NP_001026976.1 | |||
| XM_011523732.1 | XP_011522034.1 | |||
| XM_011523733.1 | XP_011522035.1 | |||
| XM_017024355.1 | XP_016879844.1 | |||
| XM_017024356.1 | XP_016879845.1 | |||
| XM_017024357.1 | XP_016879846.1 | |||
| XM_017024358.1 | XP_016879847.1 | |||
| AB208894.1 | BAD92131.1 | |||
| AK000858.1 | ||||
| AK025677.1 | ||||
| AK292381.1 | ||||
| AK315096.1 | ||||
| BC002430.2 | AAH02430.1 | |||
| CR457422.1 | CAG33703.1 | |||
| L47162.1 | AAB01003.1 | |||
| U46689.1 | AAC51121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061408 | Chr.17:19554872 - 19710044 on Build GRCh38 | Gain |
|
| nsv428337 | Chr.17:19436732 - 19682461 on Build GRCh38 | Loss |
|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
| esv3892981 | Chr.17:19585538 - 19684290 on Build GRCh38 | Gain |
|
| esv2758679 | Chr.17:19582422 - 19660453 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map