Assay Details
Target Gene Details
Entrez Gene ID: | 186 |
Gene Name: | angiotensin II receptor type 2 |
Gene Aliases: |
AT2, ATGR2, MRX88 |
Location: |
Chr.X:116170705-116174972 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGTR2 | NM_000686.4 | 3 | 1102 | NP_000677.2 |
| XM_011537533.1 | 2 | 1000 | XP_011535835.1 | |
| AK313927.1 | 3 | 1104 | ||
| BC095504.1 | 1 | 1363 | AAH95504.1 | |
| CR541969.1 | 1 | 895 | CAG46767.1 | |
| U16957.1 | 3 | 1035 | AAA67753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758886 | Chr.X:116093882 - 116232733 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map