Assay Details
Target Gene Details
Entrez Gene ID: | 7138 |
Gene Name: | troponin T1, slow skeletal type |
Gene Aliases: |
ANM, NEM5, STNT, TNT, TNTS |
Location: |
Chr.19:55132698-55149354 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNNT1 | NM_001126132.2 | 14 | 1033 | NP_001119604.1 |
| NM_001126133.2 | 13 | 1000 | NP_001119605.1 | |
| NM_001291774.1 | 14 | 1095 | NP_001278703.1 | |
| NM_003283.5 | 14 | 1081 | NP_003274.3 | |
| XM_011527246.2 | 13 | 910 | XP_011525548.1 | |
| XM_017027186.1 | 15 | 1106 | XP_016882675.1 | |
| XM_017027187.1 | 14 | 1094 | XP_016882676.1 | |
| AK291352.1 | 14 | 885 | ||
| AK308199.1 | 13 | 852 | ||
| BC010963.1 | 14 | 886 | AAH10963.1 | |
| BC022086.2 | 14 | 934 | AAH22086.2 | |
| BC034143.1 | 14 | 885 | AAH34143.1 | |
| BC107798.1 | 13 | 855 | AAI07799.1 | |
| F20363.1 | 3 | 228 | ||
| F34258.1 | 4 | 267 | ||
| M19308.1 | 13 | 853 | AAA61205.1 | |
| M19309.1 | 14 | 933 | AAA61204.1 | |
| S51938.1 | 12 | 793 | ||
| S69208.1 | 14 | 869 | AAB30272.1 | |
| S69209.1 | 13 | 836 | AAB30273.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644829 | Chr.19:55030893 - 55138732 on Build GRCh38 | Gain |
|
| nsv1060034 | Chr.19:55111224 - 55138729 on Build GRCh38 | Gain |
|
| esv2758772 | Chr.19:55026094 - 55139319 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map