Assay Details
Target Gene Details
Entrez Gene ID: | 3098 |
Gene Name: | hexokinase 1 |
Gene Aliases: |
HK, HK1-ta, HK1-tb, HK1-tc, HKD, HKI, HMSNR, HXK1, hexokinase |
Location: |
Chr.10:69270000-69401882 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HK1 | NM_001322364.1 | NP_001309293.1 | ||
| NM_001322365.1 | NP_001309294.1 | |||
| NM_033497.2 | NP_277032.1 | |||
| NM_033498.2 | NP_277033.1 | |||
| NM_033500.2 | NP_277035.2 | |||
| XM_011539732.1 | XP_011538034.1 | |||
| AK128226.1 | ||||
| DB088955.1 | ||||
| U38226.1 | AAB40527.1 | |||
| U38227.1 | AAB40528.1 | |||
| U38228.1 | AAB40529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551452 | Chr.10:69243856 - 69292752 on Build GRCh38 | Loss |
|
| nsv508589 | Chr.10:69166011 - 69276277 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map