Assay Details
Target Gene Details
Entrez Gene ID: | 958 |
Gene Name: | CD40 molecule |
Gene Aliases: |
Bp50, CDW40, TNFRSF5, p50 |
Location: |
Chr.20:46118242-46129745 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD40 | NM_001250.5 | 9 | 821 | NP_001241.1 |
| NM_001302753.1 | 9 | 861 | NP_001289682.1 | |
| NM_001322421.1 | 9 | 833 | NP_001309350.1 | |
| NM_001322422.1 | 7 | 665 | NP_001309351.1 | |
| NM_152854.3 | 8 | 759 | NP_690593.1 | |
| NR_126502.1 | 7 | 824 | ||
| NR_136327.1 | 8 | 727 | ||
| XM_005260619.3 | 7 | 689 | XP_005260676.1 | |
| XM_017028135.1 | 9 | 868 | XP_016883624.1 | |
| XM_017028136.1 | 8 | 765 | XP_016883625.1 | |
| XM_017028137.1 | 9 | 825 | XP_016883626.1 | |
| AB209660.1 | 6 | 5238 | BAD92897.1 | |
| AJ300189.1 | 8 | 697 | CAC29424.1 | |
| AK222896.1 | 9 | 761 | BAD96616.1 | |
| BC012419.1 | 9 | 788 | AAH12419.1 | |
| BC064518.1 | 7 | 800 | AAH64518.1 | |
| BT019901.1 | 9 | 731 | AAV38704.1 | |
| BX364284.2 | 1 | 809 | ||
| BX364285.2 | ||||
| KU177966.1 | 9 | 731 | ||
| KU177967.1 | 7 | 575 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057686 | Chr.20:46105657 - 46434770 on Build GRCh38 | Loss |
|
| esv2758796 | Chr.20:46064276 - 46336238 on Build GRCh38 | Loss |
|
| nsv833993 | Chr.20:46091252 - 46290015 on Build GRCh38 | Gain |
|
| nsv586135 | Chr.20:46126472 - 46168923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map