Assay Details
Target Gene Details
Entrez Gene ID: | 3784 |
Gene Name: | potassium voltage-gated channel subfamily Q member 1 |
Gene Aliases: |
ATFB1, ATFB3, JLNS1, KCNA8, KCNA9, KVLQT1, Kv1.9, Kv7.1, LQT, LQT1, RWS, SQT2, WRS |
Location: |
Chr.11:2444991-2849110 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ1 | NM_000218.2 | 16 | 2670 | NP_000209.2 |
| NM_181798.1 | 16 | 2437 | NP_861463.1 | |
| AF000571.1 | 16 | 2672 | AAC51776.1 | |
| AK129678.1 | ||||
| BC017074.1 | 11 | 1711 | AAH17074.1 | |
| U89364.1 | 16 | 2364 | AAC51781.1 |
Target Gene Details
Entrez Gene ID: | 338653 |
Gene Name: | KCNQ1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:2840135-2861569 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ1-AS1 | NR_130721.1 | |||
| AJ251642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951292 | Chr.11:2792271 - 2927870 on Build GRCh38 | Deletion |
|
| esv2759795 | Chr.11:2767814 - 2942382 on Build GRCh38 | Gain |
|
| nsv469930 | Chr.11:2831299 - 2924715 on Build GRCh38 | Loss |
|
| nsv1053066 | Chr.11:2829552 - 2872696 on Build GRCh38 | Gain |
|
| nsv553111 | Chr.11:2842057 - 2904391 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map