Assay Details
Target Gene Details
Entrez Gene ID: | 27185 |
Gene Name: | disrupted in schizophrenia 1 |
Gene Aliases: |
C1orf136, SCZD9 |
Location: |
Chr.1:231626815-232041272 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Target Gene Details
Entrez Gene ID: | 27184 |
Gene Name: | disrupted in schizophrenia 2 (non-protein coding) |
Gene Aliases: |
DISC1-AS1, DISC1OS, NCRNA00015 |
Location: |
Chr.1:231814626-231818517 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DISC2 | NR_002227.2 | 1 | 73 |
Target Gene Details
Entrez Gene ID: | 100303453 |
Gene Name: | TSNAX-DISC1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.1:231528653-232041272 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSNAX-DISC1 | NR_028393.1 | 12 | 2575 | |
NR_028394.1 | 14 | 2758 | ||
NR_028395.1 | 13 | 2655 | ||
NR_028396.1 | 13 | 2630 | ||
NR_028397.1 | 13 | 2495 | ||
NR_028398.1 | 11 | 2249 | ||
FJ804217.1 | 12 | 2575 | ||
FJ804218.1 | 13 | 2630 | ||
FJ804219.1 | 14 | 2758 | ||
FJ804220.1 | 13 | 2655 | ||
FJ804221.1 | 13 | 2495 | ||
FJ804222.1 | 11 | 2249 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762173 | Chr.1:231790208 - 231819980 on Build GRCh38 | Loss |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
