Assay Details
Target Gene Details
Entrez Gene ID: | 367 |
Gene Name: | androgen receptor |
Gene Aliases: |
AIS, AR8, DHTR, HUMARA, HYSP1, KD, NR3C4, SBMA, SMAX1, TFM |
Location: |
Chr.X:67544032-67730619 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AR | NM_000044.3 | 1 | 2408 | NP_000035.2 |
| AF162704.1 | 2 | 1275 | AAD45921.1 | |
| AF321914.1 | 1 | 1371 | AAK09423.1 | |
| AF321915.1 | 1 | 1371 | AAK09424.1 | |
| AF321916.1 | 1 | 1371 | AAK09425.1 | |
| AF321917.1 | 2 | 1347 | AAK09426.1 | |
| BC132975.1 | 1 | 1756 | ||
| FJ235916.1 | 1 | 1620 | ||
| FJ235917.1 | 1 | 1620 | ||
| FJ235918.1 | 1 | 1620 | ||
| FJ235919.1 | 1 | 1620 | ||
| FJ235920.1 | 1 | 1620 | ||
| GU208210.1 | 1 | 1296 | ||
| HM055487.1 | 1 | 1620 | ||
| KJ534771.1 | 1 | 1293 | ||
| L29496.1 | 1 | 723 | AAA51770.1 | |
| M20132.1 | 1 | 1655 | AAA51729.1 | |
| M21748.1 | 1 | 1456 | AAA51771.1 | |
| M23263.1 | 1 | 1825 | AAA51775.1 | |
| M34233.1 | 2 | 1380 | AAA51780.1 | |
| M73069.1 | 1 | 1347 | AAA51735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758871 | Chr.X:67279827 - 68099252 on Build GRCh38 | Gain |
|
| nsv818030 | Chr.X:67496364 - 68089458 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map