Assay Details
Target Gene Details
Entrez Gene ID: | 152 |
Gene Name: | adrenoceptor alpha 2C |
Gene Aliases: |
ADRA2L2, ADRA2RL2, ADRARL2, ALPHA2CAR |
Location: |
Chr.4:3766569-3768526 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRA2C | NM_000683.3 | 1 | 403 | NP_000674.2 |
| BC142625.1 | 1 | 574 | ||
| J03853.1 | 1 | 403 | AAA35513.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv998670 | Chr.4:3452060 - 3907808 on Build GRCh38 | Gain |
|
| esv26274 | Chr.4:3765714 - 3768719 on Build GRCh38 | Loss |
|
| nsv519631 | Chr.4:3752639 - 3778345 on Build GRCh38 | Loss |
|
| nsv593470 | Chr.4:3765829 - 3768071 on Build GRCh38 | Gain+Loss |
|
| esv2759219 | Chr.4:3392617 - 4349259 on Build GRCh38 | Gain |
|
| nsv997647 | Chr.4:3751070 - 3825288 on Build GRCh38 | Gain |
|
| dgv8928n54 | Chr.4:3751690 - 3768342 on Build GRCh38 | Loss |
|
| nsv949772 | Chr.4:3699474 - 3814873 on Build GRCh38 | Deletion |
|
| dgv8929n54 | Chr.4:3766386 - 3768639 on Build GRCh38 | Loss |
|
| nsv593468 | Chr.4:3758366 - 3767403 on Build GRCh38 | Loss |
|
| esv3575674 | Chr.4:3635734 - 3924606 on Build GRCh38 | Gain |
|
| dgv854n67 | Chr.4:3763999 - 3769080 on Build GRCh38 | Gain |
|
| nsv1005210 | Chr.4:3711267 - 3772212 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138457504] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map