Assay Details
Target Gene Details
Entrez Gene ID: | 148738 |
Gene Name: | hemochromatosis type 2 (juvenile) |
Gene Aliases: |
HFE2A, HJV, JH, RGMC |
Location: |
Chr.1:146017468-146021822 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HFE2 | NM_001316767.1 | 4 | 537 | NP_001303696.1 |
| NM_145277.4 | 3 | 874 | NP_660320.3 | |
| NM_202004.3 | 3 | 351 | NP_973733.1 | |
| NM_213652.3 | 2 | 314 | NP_998817.1 | |
| NM_213653.3 | 4 | 1060 | NP_998818.1 | |
| XM_005272932.1 | 4 | 991 | XP_005272989.1 | |
| AK092682.1 | 3 | 784 | BAC03944.1 | |
| AK092692.1 | 3 | 235 | BAC03947.1 | |
| AK096905.1 | 2 | 226 | BAC04890.1 | |
| AK124273.1 | 4 | 944 | BAC85823.1 | |
| AK223575.1 | 3 | 794 | ||
| AK290831.1 | 3 | 786 | ||
| AK292742.1 | 4 | 970 | ||
| AK315852.1 | 3 | 787 | ||
| AY372521.1 | 3 | 735 | AAR22390.1 | |
| BC017926.1 | 3 | 264 | AAH17926.1 | |
| BC085604.1 | 3 | 727 | AAH85604.1 | |
| DA893500.1 | 4 | 450 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005055 | Chr.1:145642988 - 148991068 on Build GRCh38 | Loss |
|
| nsv509457 | Chr.1:145948024 - 149021821 on Build GRCh38 | Insertion |
|
| nsv2699 | Chr.1:146018222 - 146052325 on Build GRCh38 | Insertion |
|
| nsv831359 | Chr.1:145983609 - 146147220 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map