Assay Details
Target Gene Details
Entrez Gene ID: | 7173 |
Gene Name: | thyroid peroxidase |
Gene Aliases: |
MSA, TDH2A, TPX |
Location: |
Chr.2:1413461-1542729 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPO | NM_000547.5 | NP_000538.3 | ||
| NM_001206744.1 | NP_001193673.1 | |||
| NM_001206745.1 | NP_001193674.1 | |||
| NM_175719.3 | NP_783650.1 | |||
| NM_175721.3 | NP_783652.1 | |||
| NM_175722.3 | NP_783653.1 | |||
| XM_011510379.2 | XP_011508681.1 | |||
| XM_011510380.2 | XP_011508682.1 | |||
| XM_011510381.2 | XP_011508683.1 | |||
| AF439430.1 | AAL74416.1 | |||
| AF533528.1 | AAN04471.1 | |||
| AK309868.1 | ||||
| BC063107.1 | AAH63107.1 | |||
| BC095448.1 | AAH95448.1 | |||
| DA958481.1 | ||||
| J02969.1 | AAA61215.1 | |||
| J02970.1 | AAA61216.1 | |||
| M17755.2 | AAA61217.2 | |||
| X17358.1 | CAA35235.1 | |||
| Y00406.1 | CAA68467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818900 | Chr.2:1422849 - 1516904 on Build GRCh38 | Gain |
|
| nsv1005443 | Chr.2:1080998 - 1747280 on Build GRCh38 | Gain |
|
| nsv468916 | Chr.2:1375171 - 1515778 on Build GRCh38 | Gain |
|
| esv3891470 | Chr.2:1005374 - 1856757 on Build GRCh38 | Gain |
|
| nsv2567 | Chr.2:1408825 - 1442337 on Build GRCh38 | Insertion |
|
| esv3589629 | Chr.2:1422992 - 1482602 on Build GRCh38 | Gain |
|
| nsv1014918 | Chr.2:772650 - 1424430 on Build GRCh38 | Gain |
|
| dgv3701n100 | Chr.2:1416577 - 1538526 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map