Assay Details
Target Gene Details
Entrez Gene ID: | 5371 |
Gene Name: | promyelocytic leukemia |
Gene Aliases: |
MYL, PP8675, RNF71, TRIM19 |
Location: |
Chr.15:73994673-74047819 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833052 | Chr.15:73958037 - 74084809 on Build GRCh38 | Gain+Loss |
|
| esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map