Assay Details
Target Gene Details
Entrez Gene ID: | 7512 |
Gene Name: | X-prolyl aminopeptidase 2 |
Gene Aliases: |
AEACEI, APP2 |
Location: |
Chr.X:129738970-129769549 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XPNPEP2 | NM_003399.5 | 1 | 211 | NP_003390.4 |
| AK300954.1 | 1 | 232 | ||
| BC126174.1 | 1 | 128 | ||
| BC143901.1 | 1 | 128 | ||
| BP270314.1 | 1 | 202 | ||
| BX386512.2 | 1 | 211 | ||
| CD617070.1 | ||||
| U90724.2 | 1 | 231 | AAB96394.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
| nsv526197 | Chr.X:129672435 - 129826254 on Build GRCh38 | Gain |
|
| esv2758893 | Chr.X:129563598 - 129830109 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map