Assay Details
Target Gene Details
Entrez Gene ID: | 4976 |
Gene Name: | OPA1, mitochondrial dynamin like GTPase |
Gene Aliases: |
BERHS, MGM1, MTDPS14, NPG, NTG, largeG |
Location: |
Chr.3:193593144-193697811 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPA1 | NM_015560.2 | NP_056375.2 | ||
| NM_130831.2 | NP_570844.1 | |||
| NM_130832.2 | NP_570845.1 | |||
| NM_130833.2 | 5 | 796 | NP_570846.1 | |
| NM_130834.2 | NP_570847.2 | |||
| NM_130835.2 | 6 | 850 | NP_570848.1 | |
| NM_130836.2 | 6 | 904 | NP_570849.2 | |
| NM_130837.2 | 7 | 958 | NP_570850.2 | |
| XM_017006500.1 | 6 | 771 | XP_016861989.1 | |
| AB011139.1 | ||||
| BC075805.1 | AAH75805.1 | |||
| DA993450.1 |
Target Gene Details
Entrez Gene ID: | 100873941 |
Gene Name: | OPA1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:193618599-193627332 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPA1-AS1 | NR_046634.1 | |||
| BG204670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759212 | Chr.3:193516111 - 193685857 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map