Assay Details
Target Gene Details
Entrez Gene ID: | 845 |
Gene Name: | calsequestrin 2 |
Gene Aliases: |
PDIB2 |
Location: |
Chr.1:115700003-115768805 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASQ2 | NM_001232.3 | 11 | 2145 | NP_001223.2 |
| AK129831.1 | 3 | 1000 | ||
| AK129891.1 | 11 | 2120 | ||
| BC022288.1 | 11 | 2054 | AAH22288.1 | |
| BM679349.1 | ||||
| D55655.1 | 11 | 1990 | BAA23494.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760632 | Chr.1:115686742 - 115742032 on Build GRCh38 | Gain |
|
| nsv831115 | Chr.1:115661250 - 115851698 on Build GRCh38 | Gain |
|
| nsv1000760 | Chr.1:115693325 - 115742032 on Build GRCh38 | Gain |
|
| nsv547585 | Chr.1:115696747 - 115745061 on Build GRCh38 | Gain |
|
| nsv524619 | Chr.1:115690789 - 115742323 on Build GRCh38 | Gain |
|
| esv27976 | Chr.1:115592696 - 116135006 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map