Assay Details
Target Gene Details
Entrez Gene ID: | 9562 |
Gene Name: | multiple inositol-polyphosphate phosphatase 1 |
Gene Aliases: |
HIPER1, MINPP2, MIPP |
Location: |
Chr.10:87502792-87553461 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MINPP1 | NM_001178117.1 | 1 | 721 | NP_001171588.1 |
| NM_004897.4 | 1 | 721 | NP_004888.2 | |
| XM_006718078.2 | 1 | 2395 | XP_006718141.1 | |
| XM_011540379.2 | 1 | 297 | XP_011538681.1 | |
| XM_017016965.1 | 1 | 2395 | XP_016872454.1 | |
| AB209819.1 | 1 | 284 | BAD93056.1 | |
| AF046914.1 | 1 | 271 | AAD02437.1 | |
| AF084943.1 | 1 | 291 | AAD09751.1 | |
| AF084944.1 | 1 | 276 | AAD09752.1 | |
| AK304624.1 | ||||
| AK312937.1 | 1 | 312 | ||
| AL050356.1 | 1 | 293 | CAB43673.1 | |
| AY358938.1 | 1 | 276 | AAQ89297.1 | |
| BC032504.1 | 1 | 273 | AAH32504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551825 | Chr.10:87497688 - 87516942 on Build GRCh38 | Gain |
|
| esv3891880 | Chr.10:87343291 - 87725505 on Build GRCh38 | Gain |
|
| nsv428236 | Chr.10:86866859 - 87550005 on Build GRCh38 | Loss |
|
| esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss |
|
| esv26704 | Chr.10:87388736 - 87516801 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map